Learn More
Julabo USA Inc Recirculating Chiller, VALEGRO 1001H, 230V/60Hz, -20 to +85 C, 1kW cooling power, 2kW heater. OLED UI with RS232 and USB-C ports. Adjustable pump to 16PSI, 23L/min. 4-7L tank.
SDP
Catalog No. NC3907383
Encompass_Preferred
VALEGRO 1001H Chiller, 200-230V/50-60Hz (EU/US plugs), -20…+85 C ±0.2 C, R290 refrigerant; 2kW heater Cooling: 1000W @ 20C; 750W @ 0C; 500W @ -10C, 280W @ -20C. OLED touchscreen UI. Adjustable pump up to 23L/min, 16PSI. RS232 and USB-C for PC control. Optional Ethernet (activate with p/n 30004147). M16x1 fittings 8 /12mm barbs. 4-7L bath tank. 14.4 x 20.7 x 24 in, 95lbs
Catalog No. NC3907383 Supplier Julabo USA Inc Supplier No. 9612101.30005663
VALEGRO 1001H Chiller, 200-230V/50-60Hz (EU/US plugs), -20…+85 C ±0.2 C, R290 refrigerant; 2kW heater Cooling: 1000W @ 20C; 750W @ 0C; 500W @ -10C, 280W @ -20C. OLED touchscreen UI. Adjustable pump up to 23L/min, 16PSI. RS232 and USB-C for PC control. Optional Ethernet (activate with p/n 30004147). M16x1 fittings 8 /12mm barbs. 4-7L bath tank. 14.4 x 20.7 x 24 in, 95lbs