Learn More
Julabo USA Inc Recirculating Chiller, VALEGRO 1501H, 115V/60Hz, -20 to +85 C, 1.5kW cooling power, 1kW heater. OLED UI with RS232 and USB-C ports. Adjustable pump to 16PSI, 23L/min. 4-7L tank.
SDP
Catalog No. NC3907396
Encompass_Preferred
VALEGRO 1501H Chiller, 115V/60Hz (NEMA N5-15 plug), -20…+85 C ±0.2 C, R290 refrigerant; 1kW heater Cooling: 1500W @ 20C; 1000W @ 0C; 700W @ -10C, 400W @ -20C. OLED touchscreen UI. Adjustable pump 15 - 23L/min, 7.3 - 16PSI. RS232 and USB-C for PC control. Optional Ethernet (activate with p/n 30004147). M16x1 fittings 8 /12mm barbs. 4-7L bath tank. 14.4 x 20.7 x 24 in, 95lbs
Catalog No. NC3907396 Supplier Julabo USA Inc Supplier No. 9612151.30005682
VALEGRO 1501H Chiller, 115V/60Hz (NEMA N5-15 plug), -20…+85 C ±0.2 C, R290 refrigerant; 1kW heater Cooling: 1500W @ 20C; 1000W @ 0C; 700W @ -10C, 400W @ -20C. OLED touchscreen UI. Adjustable pump 15 - 23L/min, 7.3 - 16PSI. RS232 and USB-C for PC control. Optional Ethernet (activate with p/n 30004147). M16x1 fittings 8 /12mm barbs. 4-7L bath tank. 14.4 x 20.7 x 24 in, 95lbs