Learn More
Julabo USA Inc Recirculating Chiller, VALEGRO 2503, 230V/60Hz, -20 to +40 C, 2.5kW cooling power. OLED UI with RS232 and USB-C ports. Adjustable pump to 50.8PSI, 40L/min. 3-8L tank.
SDP
Catalog No. NC3907375
Encompass_Preferred
VALEGRO 2503 Chiller, 200-230V/50-60Hz (EU/US plugs), -20…+40 C ±0.3 C, R290 refrigerant; Cooling: 2500W @ 20C; 1450W @ 0C; 1000W @ -10C, 500W @ -20C. OLED touchscreen UI. Adjustable pump 15 - 40L/min, 7.3 - 50.8PSI. RS232 and USB-C for PC control. Optional Ethernet (activate with p/n 30004147). M24x1.5 fittings with 12mm hse barbs. 3-8L bath tank. 16.9 x 22 x 27.2 in, 134lbs
Catalog No. NC3907375 Supplier Julabo USA Inc Supplier No. 9610253.30006998
VALEGRO 2503 Chiller, 200-230V/50-60Hz (EU/US plugs), -20…+40 C ±0.3 C, R290 refrigerant; Cooling: 2500W @ 20C; 1450W @ 0C; 1000W @ -10C, 500W @ -20C. OLED touchscreen UI. Adjustable pump 15 - 40L/min, 7.3 - 50.8PSI. RS232 and USB-C for PC control. Optional Ethernet (activate with p/n 30004147). M24x1.5 fittings with 12mm hse barbs. 3-8L bath tank. 16.9 x 22 x 27.2 in, 134lbs