Learn More
Julabo USA Inc Recirculating Chiller, VALEGRO 2503H, 230V/60Hz, -20 to +85 C, 2.5kW cooling power, 2kW heater. OLED UI with RS232 and USB-C ports. Adjustable pump to 50.8PSI, 40L/min. 3-8L tank.
SDP
Catalog No. NC3907402
Encompass_Preferred
VALEGRO 2503H Chiller, 200-230V/50-60Hz (EU/US plugs), -20…+85 C ±0.2 C, R290 refrigerant; 2kW heater Cooling: 2500W @ 20C; 1450W @ 0C; 1000W @ -10C, 500W @ -20C. OLED touchscreen UI. Adjustable pump up to 15 - 40L/min, 7.3 - 50.8PSI. RS232 and USB-C for PC control. Optional Ethernet (activate with p/n 30004147). M16x1 fittings 8 /12mm barbs. 3-8L bath tank. 16.9 x 19.7 x 27.2 in, 134lbs
Catalog No. NC3907402 Supplier Julabo USA Inc Supplier No. 9612253.30007023
VALEGRO 2503H Chiller, 200-230V/50-60Hz (EU/US plugs), -20…+85 C ±0.2 C, R290 refrigerant; 2kW heater Cooling: 2500W @ 20C; 1450W @ 0C; 1000W @ -10C, 500W @ -20C. OLED touchscreen UI. Adjustable pump up to 15 - 40L/min, 7.3 - 50.8PSI. RS232 and USB-C for PC control. Optional Ethernet (activate with p/n 30004147). M16x1 fittings 8 /12mm barbs. 3-8L bath tank. 16.9 x 19.7 x 27.2 in, 134lbs