Learn More
LGC GENOMICS LLC (Lucigen) Rec J Exonuclease, 250 U @ 10 U/LMagnesium-dependent exonuclease for 5' to 3' digestion of single-stranded DNA.
Catalog No. NC1058432
Encompass
Rec J Exonuclease, derived from E. coli, catalyzes degradation of ssDNA in a 53 direction. Its activity is dependent on Mg2+. Rec J Exonuclease can be heat-inactivated by incubation at 65C for 20 minutes.
This item is not returnable. View return policy
Catalog No. NC1058432 Supplier LGC GENOMICS LLC Supplier No. RJ411250
Rec J Exonuclease, derived from E. coli, catalyzes degradation of ssDNA in a 53 direction. Its activity is dependent on Mg2+. Rec J Exonuclease can be heat-inactivated by incubation at 65C for 20 minutes.