Learn More
Aviva Systems Biology Corp Reck Antibody - middle region (ARP59228_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59228_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Reck. It was validated on Western Blot. . Alternate names: MGC169689, St15, mRECKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-624