Learn More
Medicus Health Reclining Power Adjustment Phlebotomy Chair
SDP
Catalog No. NC2230121
Encompass
Phlebotomy Chair Offers Smooth, Fully Electric Reclining and Height Adjustment keeps patients comfortable and may better accommodate patients who may faint during a procedure ∙ Self leveling arms and synchronized footrest that move with chair's seat and backrest ∙ Keeps patients comfortable and may better accommodate patients who may faint during a procedure ∙ Power supplies and motors meet UL 60601-1 and CAN/CSA C22.2 601.1 ∙ Rated 5 amps at 120 VAC ∙ Comes with hand controller All-welded, steel, neutral gray powder-coated frame ∙ Padded seat, backrest, armrests and footrest ∙ Armrests flip up out of the way for easy entry and exit ∙ Adjustable foot levelers ∙ Easy-clean, durable, stain-resistant vinyl∙ 375 lbs. load capacity under normal use (170 kg) ∙ Chairs are not engineered to restrain a fainting or falling occupant. The chair’s occupant should NEVER be left unattended
This item is not returnable. View return policy
Catalog No. NC2230121 Supplier Medicus Health Supplier No. 2666M1WEDGEWOOD
Phlebotomy Chair Offers Smooth, Fully Electric Reclining and Height Adjustment keeps patients comfortable and may better accommodate patients who may faint during a procedure ∙ Self leveling arms and synchronized footrest that move with chair's seat and backrest ∙ Keeps patients comfortable and may better accommodate patients who may faint during a procedure ∙ Power supplies and motors meet UL 60601-1 and CAN/CSA C22.2 601.1 ∙ Rated 5 amps at 120 VAC ∙ Comes with hand controller All-welded, steel, neutral gray powder-coated frame ∙ Padded seat, backrest, armrests and footrest ∙ Armrests flip up out of the way for easy entry and exit ∙ Adjustable foot levelers ∙ Easy-clean, durable, stain-resistant vinyl∙ 375 lbs. load capacity under normal use (170 kg) ∙ Chairs are not engineered to restrain a fainting or falling occupant. The chair’s occupant should NEVER be left unattended