Learn More
Marketlab Reclining Power Blood Draw Chair
Catalog No. 22505149
Change view
Click to view available options
Color:
Beige
Black
Burgundy
Gunmetal Gray
Light Gray
Royal Blue
Slate Blue
Warm Gray
Wedgewood Blue
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-505-149 Wedgewood Blue
22-505-085 Black
22-505-135 Light Gray
22-505-076 Beige
22-505-099 Gunmetal Gray
22-505-136 Royal Blue
22-505-128 Slate Blue
22-505-101 Warm Gray
22-505-118 Burgundy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 22-505-149 Supplier Marketlab Supplier No. 15995WD
Add to Cart
Add to Cart
Reclining Power Draw Chair with electric recline, hand controller, and CAL 117 Vinyl upholstery.
  • 35 in. W x 35 in. D x 50 in. H chair with CAL 117 vinyl upholstery.
  • New soft, curved armrest and seat edges for improved comfort and durability.
  • Upholstery stitching process with fewer crevices for enhanced infection control.
  • Waterfall seat edge.
  • 110-volt lift mechanism enclosed in smart-locking, brushed aluminum casing.
  • Hand controller.
  • Smooth electric 30° recline.
  • Chair length range is 35 in. when upright to 66 in. when fully laid back; seat height adjustment range is 22 to 30 in.; back: 20 in. W x 32 in. H 25-75 degree angle; footrest: 19.5 in. W x 14 in. D 5-90 degree angle.
  • 375 lb. weight capacity.
Specifications
Product Type Powered Phlebotomy Chair
Arm Style Straight Arm
Color Wedgewood Blue
Material Vinyl Upholstery