Learn More
Uline 5 x 5" 2 Mil Reclosable Bags 1,000/cs
Catalog No. NC3235604
Encompass_Preferred
The prices are right on these quality reclosable poly bags.Single-track zipper closure keeps out dirt and moisture.All virgin polyethylene film meets FDA and USDA requirements.
Catalog No. NC3235604 Supplier Uline Supplier No. S1899
The prices are right on these quality reclosable poly bags.Single-track zipper closure keeps out dirt and moisture.All virgin polyethylene film meets FDA and USDA requirements.