Learn More
Grainger Reclosable Fastener 1 x 3 In PK100
Catalog No. 502765356
Encompass_Preferred
The Reclosable Fastener, 1" x 3" from Grainger is a pack of 100 plastic, black cable ties that have a one-time lock feature for securing bundles and attaching items. They have a minimum loop tensile strength of 50 lb and a maximum bundle diameter of 2.75". The operating temperature range is -40°F to 185°F. The cable tie heads are 0.1875"H x 0.5"W with a strap thickness of 0.05" and strap width of 0.1875". This product can be used for bundling wires, cables, and tubing.
Catalog No. 50-276-5356 Supplier Grainger Supplier No. 16U707
The Reclosable Fastener, 1" x 3" from Grainger is a pack of 100 plastic, black cable ties that have a one-time lock feature for securing bundles and attaching items. They have a minimum loop tensile strength of 50 lb and a maximum bundle diameter of 2.75". The operating temperature range is -40°F to 185°F. The cable tie heads are 0.1875"H x 0.5"W with a strap thickness of 0.05" and strap width of 0.1875". This product can be used for bundling wires, cables, and tubing.