Learn More
Grainger Reclosable Fastener 1x2 In PK100
Catalog No. 502766754
Encompass_Preferred
The 3M Dual Lock Hook and Loop Fastener Strip is a reclosable fastener that creates a strong, secure hold. It features a continuous mushroom-shaped stem that engages with a mating piece to create a strong bond. This fastener strip has an adhesive backing for easy application and can be cut to desired lengths. It can withstand repeated closures, is water resistant, and bonds on contact without heat or solvents. This versatile fastener is ideal for attaching, mounting or fastening items for industrial, commercial and consumer projects.
Catalog No. 50-276-6754 Supplier Grainger Supplier No. 16U709
The 3M Dual Lock Hook and Loop Fastener Strip is a reclosable fastener that creates a strong, secure hold. It features a continuous mushroom-shaped stem that engages with a mating piece to create a strong bond. This fastener strip has an adhesive backing for easy application and can be cut to desired lengths. It can withstand repeated closures, is water resistant, and bonds on contact without heat or solvents. This versatile fastener is ideal for attaching, mounting or fastening items for industrial, commercial and consumer projects.