Learn More
Grainger Reclosable Fastener Black Low Profile
Catalog No. 502768398
Encompass_Preferred
The Tripp Lite 16U584 is a 19-inch open frame rack enclosure cabinet with adjustable mounting rails. It has an overall height of 34 inches, width of 24 inches, and depth of 24 inches. The steel enclosure features a vented top panel, split rear door, and side panels with quick-release latches. The front and rear doors are removable and reversible. The adjustable mounting rails have square mounting holes and numbered rack spaces. This rack provides 84U of storage space for mounting rackmount equipment. The open frame design allows for efficient cooling.
Catalog No. 50-276-8398 Supplier Grainger Supplier No. 16U584
The Tripp Lite 16U584 is a 19-inch open frame rack enclosure cabinet with adjustable mounting rails. It has an overall height of 34 inches, width of 24 inches, and depth of 24 inches. The steel enclosure features a vented top panel, split rear door, and side panels with quick-release latches. The front and rear doors are removable and reversible. The adjustable mounting rails have square mounting holes and numbered rack spaces. This rack provides 84U of storage space for mounting rackmount equipment. The open frame design allows for efficient cooling.