Learn More
Grainger Reclosable Fastener Clear Low Profile
Catalog No. 502767663
Encompass_Preferred
The Grainger 16U583 is a wall-mounted network cabinet that provides secure storage for networking and telecom equipment. This steel cabinet measures 24 inches wide x 24 inches deep x 42 inches high and comes in a black powder coat finish. It features locking, removable side panels and front/rear doors with vented sides for airflow. The cabinet includes mounting rails and has a weight capacity of 200 lbs. Overall, this network cabinet offers sturdy, enclosed storage to protect rack-mounted equipment in office and data center environments.
Catalog No. 50-276-7663 Supplier Grainger Supplier No. 16U583
The Grainger 16U583 is a wall-mounted network cabinet that provides secure storage for networking and telecom equipment. This steel cabinet measures 24 inches wide x 24 inches deep x 42 inches high and comes in a black powder coat finish. It features locking, removable side panels and front/rear doors with vented sides for airflow. The cabinet includes mounting rails and has a weight capacity of 200 lbs. Overall, this network cabinet offers sturdy, enclosed storage to protect rack-mounted equipment in office and data center environments.