Learn More
Grainger Reclosable Fastener Dual Lock 2in PK100
Catalog No. 502765045
Encompass_Preferred
The 3M Dual Lock Hook and Loop Fastener Strip is a heavy duty reclosable fastener that creates a strong, secure bond. It features a mushroom-shaped stem design that interlocks with a mated piece to provide high strength and holding power. This fastener strip has an adhesive backing for easy application and can bond to a variety of surfaces including plastic, metal, wood, and more. It is available in black, white or clear and in multiple widths and lengths. This versatile fastener is ideal for mounting, attaching and assembling items for industrial, commercial and consumer projects.
Catalog No. 50-276-5045 Supplier Grainger Supplier No. 16U597
The 3M Dual Lock Hook and Loop Fastener Strip is a heavy duty reclosable fastener that creates a strong, secure bond. It features a mushroom-shaped stem design that interlocks with a mated piece to provide high strength and holding power. This fastener strip has an adhesive backing for easy application and can bond to a variety of surfaces including plastic, metal, wood, and more. It is available in black, white or clear and in multiple widths and lengths. This versatile fastener is ideal for mounting, attaching and assembling items for industrial, commercial and consumer projects.