Learn More
Grainger Reclosable Fastener Dual Lock Clear
Catalog No. 502768333
Encompass_Preferred
The 3M Dual Lock Hook and Loop Fastener Roll is a heavy duty reclosable fastener that creates a strong, secure bond. It has a mushroom-shaped stem that interlocks with another piece of Dual Lock fastener to form a connection. This product has an adhesive backing for attachment to surfaces. It is available in a 1" x 10 yard roll size. The fastener is made from nylon and is good for indoor and outdoor use. It can withstand vibration, shock, and harsh conditions. The Dual Lock fastener is versatile and can bond a wide range of materials including plastic, metal, glass, wood, and more. It provides a quick attach and detach capability.
Catalog No. 50-276-8333 Supplier Grainger Supplier No. 16U580
The 3M Dual Lock Hook and Loop Fastener Roll is a heavy duty reclosable fastener that creates a strong, secure bond. It has a mushroom-shaped stem that interlocks with another piece of Dual Lock fastener to form a connection. This product has an adhesive backing for attachment to surfaces. It is available in a 1" x 10 yard roll size. The fastener is made from nylon and is good for indoor and outdoor use. It can withstand vibration, shock, and harsh conditions. The Dual Lock fastener is versatile and can bond a wide range of materials including plastic, metal, glass, wood, and more. It provides a quick attach and detach capability.