Learn More
Grainger Reclosable Fastener Dual Lock 3in PK100
Catalog No. 502764906
Encompass_Preferred
The 3M PK-100 3-Ply Disposable Face Masks are designed to help provide respiratory protection for the wearer. They feature a 3-ply construction and have a fluid resistant outer facing that provides protection against large droplets, sprays, or splashes that may contain germs. The masks have a soft inner facing for comfort against the face. The masks feature dual elastic headbands to provide a custom and secure seal. They are latex-free and are intended for single use only. The masks meet ASTM F2100-19 Level 1 standards and are FDA cleared for use as surgical masks. They provide >95% BFE and >95% PFE based on independent testing. Each box contains 100 disposable face masks.
Catalog No. 50-276-4906 Supplier Grainger Supplier No. 16U605
The 3M PK-100 3-Ply Disposable Face Masks are designed to help provide respiratory protection for the wearer. They feature a 3-ply construction and have a fluid resistant outer facing that provides protection against large droplets, sprays, or splashes that may contain germs. The masks have a soft inner facing for comfort against the face. The masks feature dual elastic headbands to provide a custom and secure seal. They are latex-free and are intended for single use only. The masks meet ASTM F2100-19 Level 1 standards and are FDA cleared for use as surgical masks. They provide >95% BFE and >95% PFE based on independent testing. Each box contains 100 disposable face masks.