Learn More
Grainger Reclosable Fastener Hook 15 ft.
Catalog No. 502766644
Encompass_Preferred
The Reclosable Fastener Hook from 3M is a plastic hook fastener that can be opened and closed repeatedly for easy fastening and unfastening. It has an adhesive backing for attachment to smooth surfaces. The hook side fastener is 2 inches wide by 2 inches long. It can withstand temperatures up to 176°F. The adhesive provides a strong, long-lasting bond. This reclosable fastener hook can be used for attaching signs, holding wires and cables, making adjustable straps, and various other fastening applications. It is sold individually.
Catalog No. 50-276-6644 Supplier Grainger Supplier No. 16U607
The Reclosable Fastener Hook from 3M is a plastic hook fastener that can be opened and closed repeatedly for easy fastening and unfastening. It has an adhesive backing for attachment to smooth surfaces. The hook side fastener is 2 inches wide by 2 inches long. It can withstand temperatures up to 176°F. The adhesive provides a strong, long-lasting bond. This reclosable fastener hook can be used for attaching signs, holding wires and cables, making adjustable straps, and various other fastening applications. It is sold individually.