Learn More
Grainger Reclosable Fastener Loop 15 ft.
Catalog No. 502766743
Encompass_Preferred
The 3M Dual Lock Hook and Loop Fastener Strip is a reclosable fastener that creates a strong, secure hold. It features a continuous mushroom-shaped stem that engages with a mating piece to create a strong bond. This fastener strip has an adhesive backing for easy application and can be cut to desired lengths. It can withstand repeated closures, is water resistant, and bonds on contact with no need for pressure. This versatile fastener is ideal for mounting, attaching, and assembling items.
Catalog No. 50-276-6743 Supplier Grainger Supplier No. 16U608
The 3M Dual Lock Hook and Loop Fastener Strip is a reclosable fastener that creates a strong, secure hold. It features a continuous mushroom-shaped stem that engages with a mating piece to create a strong bond. This fastener strip has an adhesive backing for easy application and can be cut to desired lengths. It can withstand repeated closures, is water resistant, and bonds on contact with no need for pressure. This versatile fastener is ideal for mounting, attaching, and assembling items.