Learn More
Grainger Reclosable Fastener Loop 30 ft.
Catalog No. 502767715
Encompass_Preferred
The 3M Reclosable Fastener Roll is a versatile hook and loop fastener tape that can be cut to size. It features a strong adhesive backing for secure attachment to fabric, plastic, metal, wood, and more. This industrial strength fastener has a width of 1" and comes in a 25 yard roll. It can withstand repeated closures and is washable. The hook side is black while the loop side is white. This product can be used for a wide variety of DIY, crafting, and industrial applications requiring a secure, reusable closure.
Catalog No. 50-276-7715 Supplier Grainger Supplier No. 16U575
The 3M Reclosable Fastener Roll is a versatile hook and loop fastener tape that can be cut to size. It features a strong adhesive backing for secure attachment to fabric, plastic, metal, wood, and more. This industrial strength fastener has a width of 1" and comes in a 25 yard roll. It can withstand repeated closures and is washable. The hook side is black while the loop side is white. This product can be used for a wide variety of DIY, crafting, and industrial applications requiring a secure, reusable closure.