Learn More
Grainger 3M DUAL LOCK Reclosable Fastener VHB Acrylic
Catalog No. NC1937610
Encompass_Preferred
The 3M Dual Lock Reclosable Fastener VHB Acrylic Foam Tape is an acrylic foam tape with reclosable fasteners on both sides. It has a high-strength adhesive and can bond two rigid surfaces together to create a quick attachment system. This VHB tape can withstand dynamic loads and shear forces while still allowing parts to be removed and reattached. It has a service temperature range of -40°F to 200°F and comes in multiple sizes and thicknesses. This is a versatile fastening solution for industrial, electronic, automotive, and construction applications.
Catalog No. NC1937610 Supplier Grainger Supplier No. 454W99
The 3M Dual Lock Reclosable Fastener VHB Acrylic Foam Tape is an acrylic foam tape with reclosable fasteners on both sides. It has a high-strength adhesive and can bond two rigid surfaces together to create a quick attachment system. This VHB tape can withstand dynamic loads and shear forces while still allowing parts to be removed and reattached. It has a service temperature range of -40°F to 200°F and comes in multiple sizes and thicknesses. This is a versatile fastening solution for industrial, electronic, automotive, and construction applications.