Learn More
Alkali Scientific Reclosable Specimen Bags with Zip Lock Closure and Marking Area, LDPE, 40 µm Thickness, 120x170mm, 6,000/Case
SDP
Catalog No. 5000663177
Encompass_Preferred
These reclosable specimen bags are manufactured from durable LDPE (low density polyethylene) and are designed for the secure containment, organization, and handling of specimens and samples in laboratory, clinical, and research environments. Each bag features a reliable zip lock closure that allows easy opening and resealing while helping maintain contents securely during handling and storage. A dedicated marking area supports clear labeling and sample identification. With a 40 µm material thickness, the bags provide an effective balance of flexibility and strength for routine specimen handling. Available in multiple s, they accommodate small to larger samples and are commonly used for storage, sorting, internal transport, and general laboratory workflows.
Catalog No. 50-006-63177 Supplier Alkali Scientific Supplier No. SB70120X170
These reclosable specimen bags are manufactured from durable LDPE (low density polyethylene) and are designed for the secure containment, organization, and handling of specimens and samples in laboratory, clinical, and research environments. Each bag features a reliable zip lock closure that allows easy opening and resealing while helping maintain contents securely during handling and storage. A dedicated marking area supports clear labeling and sample identification. With a 40 µm material thickness, the bags provide an effective balance of flexibility and strength for routine specimen handling. Available in multiple s, they accommodate small to larger samples and are commonly used for storage, sorting, internal transport, and general laboratory workflows.