Learn More
Vector Laboratories Recombinant Ebola Virus VP35 Protein [6C5], Mouse IgG2a, Kappa
SDP
Catalog No. NC2744063
Encompass
This mouse IgG2a monoclonal antibody [6C5] was generated against DNA immunization followed by recombinant protein and is specific for the C-terminal portion of Zaire ebolavirus polymerase cofacter VP35 protein.Highlights:Reacts with the C-terminal portion of Zaire ebolavirus polymerase cofacter VP35 proteinSuitable for Western Blot, Immunoprecipitation and Immunofluorescence applications
Catalog No. NC2744063 Supplier Vector Laboratories Supplier No. KFAB012292.0
This mouse IgG2a monoclonal antibody [6C5] was generated against DNA immunization followed by recombinant protein and is specific for the C-terminal portion of Zaire ebolavirus polymerase cofacter VP35 protein.Highlights:Reacts with the C-terminal portion of Zaire ebolavirus polymerase cofacter VP35 proteinSuitable for Western Blot, Immunoprecipitation and Immunofluorescence applications