Learn More
Abcam Recombinant human CDC42 protein (Active), 5UG
Supplier:  Abcam AB2683905UG
Encompass_Preferred
Recombinant human CDC42 protein (Active) is a Human Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Cell division control protein 42 homolog, G25K GTP-binding protein, CDC42
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2455064