Learn More
Abcam Recombinant human JAK1 protein (Active), 10UG
Supplier:  Abcam AB21619410UG
Encompass_Preferred
Recombinant human JAK1 protein (Active) is a Human Fragment protein, in the 866 to 1154 aa range, expressed in Baculovirus infected Sf9, with >=35% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: JAK1A, JAK1B, JAK1, Tyrosine-protein kinase JAK1, Janus kinase 1, JAK-1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2207956