Learn More
Creative Biolabs RECMB MOU ANTI-NGFR AB 300UG
SDP
Catalog No. NC2873827
Encompass
Recombinant Mouse Anti-NGFR Antibody (5H1) 300ugConjugation: NONEPurity: SDS-PAGE>95%
Catalog No. NC2873827 Supplier Creative Biolabs Supplier No. NEUT1779CQ300UG
Recombinant Mouse Anti-NGFR Antibody (5H1) 300ugConjugation: NONEPurity: SDS-PAGE>95%