Learn More
Worthington Biochemical Corporation RECMB SERRATIA MARC-NUCL100KU
SDP
Catalog No. NC3672529
Encompass
Recombinant Serratia marcescens nuclease produced in Komagataella phaffi. Chromatographically purified. Free of animal derived components and proteases. A liquid preparation in 20mM Tris-HCl, 2mM MgCl2, 20mM NaCl, pH 8.0 and 50% glycerol. Store at -20°C. 100 KU
Catalog No. NC3672529 Supplier Worthington Biochemical Corporation Supplier No. LS01524
Recombinant Serratia marcescens nuclease produced in Komagataella phaffi. Chromatographically purified. Free of animal derived components and proteases. A liquid preparation in 20mM Tris-HCl, 2mM MgCl2, 20mM NaCl, pH 8.0 and 50% glycerol. Store at -20°C. 100 KU