Learn More
Medchemexpress LLC RecO E.coli His-SU 50ug
SDP
Catalog No. 5000195904
Encompass_Preferred
RecO Protein, E. coli (His-SUMO) is a recombinant Escherichia coli DNA repair protein supplied as a lyophilized preparation with an N-terminal His-SUMO fusion tag. It is intended for research use in DNA recombination and repair studies; consult the Certificate of Analysis for lot-specific concentration and storage instructions.
  • Recombinant E. coli RecO with N-terminal His-SUMO tag for purification and detection.
  • Lyophilized powder for improved stability and long-term storage.
  • Approximate molecular weight 43.4 kDa for experimental planning.
  • Suitable for DNA repair and recombination studies.
  • Certificate of Analysis and SDS available for lot-specific details.
Catalog No. 50-001-95904 Supplier Medchemexpress LLC Supplier No. HYP7151250UG
RecO Protein, E. coli (His-SUMO) is a recombinant Escherichia coli DNA repair protein supplied as a lyophilized preparation with an N-terminal His-SUMO fusion tag. It is intended for research use in DNA recombination and repair studies; consult the Certificate of Analysis for lot-specific concentration and storage instructions.
  • Recombinant E. coli RecO with N-terminal His-SUMO tag for purification and detection.
  • Lyophilized powder for improved stability and long-term storage.
  • Approximate molecular weight 43.4 kDa for experimental planning.
  • Suitable for DNA repair and recombination studies.
  • Certificate of Analysis and SDS available for lot-specific details.