Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RecO E.coli His-SU 5ug
SDP

Catalog No. 5000196395
Encompass_Preferred

Recombinant RecO protein from Escherichia coli expressed with an N-terminal His-SUMO affinity tag, supplied lyophilized for biochemical and molecular biology studies of DNA recombination and repair. The product is formulated for stability and quality and is intended for binding, enzymology, and structural assays after reconstitution.

  • Contains an N-terminal His-SUMO affinity tag for straightforward purification and detection.
  • Purified to greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight 43.4 kDa, suitable for biochemical characterization.
  • Lyophilized from 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0, to enhance stability.
  • Recommended storage at -20°C or -80°C; stable at 4°C for one week after reconstitution.
  • Available in small, research-friendly sizes to facilitate assay optimization.

Catalog No. 50-001-96395 Supplier Medchemexpress LLC Supplier No. HYP715125UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant RecO protein from Escherichia coli expressed with an N-terminal His-SUMO affinity tag, supplied lyophilized for biochemical and molecular biology studies of DNA recombination and repair. The product is formulated for stability and quality and is intended for binding, enzymology, and structural assays after reconstitution.

  • Contains an N-terminal His-SUMO affinity tag for straightforward purification and detection.
  • Purified to greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight 43.4 kDa, suitable for biochemical characterization.
  • Lyophilized from 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0, to enhance stability.
  • Recommended storage at -20°C or -80°C; stable at 4°C for one week after reconstitution.
  • Available in small, research-friendly sizes to facilitate assay optimization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.