Learn More
Abcam Recombinant Anti-TTF1 antibody [EPR5955(2)]Produced recombinantly (animal-free) for high batch-to-batch consistency and long term security of supplyRabbit monoclonal [EPR5955(2)] to TTF1Suitable for: WB, IHC-PReacts with: Mouse, Rat, Human
Supplier:  Abcam AB133737100UL
Encompass_Preferred
Recombinant Anti-TTF1 antibody [EPR5955(2)]Produced recombinantly (animal-free) for high batch-to-batch consistency and long term security of supplyRabbit monoclonal [EPR5955(2)] to TTF1Suitable for: WB, IHC-PReacts with: Mouse, Rat, Human
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2092321