Learn More
Abcam Recombinant human JAK2 protein, 20UG
Supplier:  Abcam AB4261920UG
Encompass_Preferred
Recombinant human JAK2 protein is a Human Fragment protein, in the 808 to 1132 aa range, expressed in Baculovirus infected Sf9, with >70% purity and suitable for SDS-PAGE, FuncS, Inhib.
Protein/ Peptide/ Enzyme Synonyms: Tyrosine-protein kinase JAK2, Janus kinase 2, JAK-2, JAK2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2065948