Learn More
Abcam Recombinant human PLK1 protein, 100UG
Supplier:  Abcam AB51426100UG
Encompass_Preferred
Recombinant human PLK1 protein is a Human Full Length protein, in the 1 to 603 aa range, expressed in Baculovirus infected Sf9, with >70% purity and suitable for WB, FuncS, SDS-PAGE, IHC-P.
Protein/ Peptide/ Enzyme Synonyms: PLK, PLK1, Serine/threonine-protein kinase PLK1, Polo-like kinase 1, Serine/threonine-protein kinase 13, PLK-1, STPK13
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1696986