Learn More
Abcam Recombinant human TNF alpha protein, 10UG
Supplier:  Abcam AB964210UG
Encompass_Preferred
Recombinant human TNF alpha protein is a Human Full Length protein, in the 77 to 233 aa range with >98% purity, < 1 EU/µg endotoxin level and suitable for ELISA, SDS-PAGE, Functional studies and HPLC. The predicted molecular weight of ab9642 protein is 17.4 kDa. - Save time and ensure accurate results - use our TNF alpha protein as a control - Optimal protein bioactivity and stability - Available in different sizes to fit your experimental needs - Cited in over 25 publications
Protein/ Peptide/ Enzyme Synonyms: TNFA, TNFSF2, TNF, Tumor necrosis factor, Cachectin, TNF-alpha, Tumor necrosis factor ligand superfamily member 2, TNF-a
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1475094