Learn More
Abcam Recombinant mouse Hsp70 protein, 50UG
Supplier:  Abcam AB11318750UG
Encompass_Preferred
Recombinant mouse Hsp70 protein is a Mouse Full Length protein, in the 1 to 641 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE, WB, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Hsp70-3, Hsp70A1, Hspa1a, Heat shock 70 kDa protein 1A, Heat shock 70 kDa protein 3, Hsp68, HSP70.3
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2006820