Learn More
Eurofins Discoverx Recombinant Human soluble RANK ligand, 10 µg. (92-1282)
Supplier:  Eurofins Discoverx 921282
Encompass
Soluble RANK ligand (sRANKL) is a type II protein and member of the tumor necrosis factor (TNF) superfamily which serves as an agonist for RANK. Controls bone regeneration and remodeling, and affects the immune system. Key factor in osteoclast differentiation and activation, dendritic cell survival, and T cell-dependent immune response. Provided as 10 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9799