Learn More
Abcam Recombinant Monoclonal Antibody, Alexa Fluor 488, 0.5 mg/mL, Mouse reactivity, Clone EPR17067
Supplier:  Abcam AB208271100UL
Encompass_Preferred
This product is a recombinant rabbit monoclonal antibody that targets Interferon Regulatory Factor 5 (IRF5) and is conjugated with Alexa Fluor 488. IRF5 is a crucial transcription factor found in immune cells, modulating the immune response and influencing the production of pro-inflammatory cytokines. This antibody aids in understanding immune function and responses during viral infections due to its involvement in TLR signaling pathways.
  • Produced using recombinant monoclonal technology for high batch-to-batch consistency and reproducibility.
  • Directly conjugated to Alexa Fluor 488, eliminating the need for secondary antibodies and reducing experimental complexity.
  • Enables specific detection of IRF5, important for studying cytokine production and antiviral defense.
  • Validated for use in immunocytochemistry/immunofluorescence, ensuring reliable performance.
Catalog No. NC2268895