Learn More
Abcam Produced recombinantly (animal-free) for high batch-to-batch consistency and long term security of supply.Rabbit monoclonal [EPR5145(2)] to SPT5,Suitable for: WB, IHC-P, ICC/IF,Reacts with: Mouse, Rat, Human
Supplier:  Abcam AB126592100UL
Encompass_Preferred
Produced recombinantly (animal-free) for high batch-to-batch consistency and long term security of supply.Rabbit monoclonal [EPR5145(2)] to SPT5,Suitable for: WB, IHC-P, ICC/IF,Reacts with: Mouse, Rat, Human
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2874126