Learn More
Abcam RECOMB ANTI-WISP1 ANTBDY,100UG
Supplier:  Abcam AB281140100UG
Encompass_Preferred
Recombinant Anti-WISP1 antibody [EPR22891-92] - BSA and Azide free (Capture) (ab281140)Produced recombinantly (animal-free) for high batch-to-batch consistency and long term security of supplyRabbit monoclonal [EPR22891-92] to WISP1 - BSA and Azide free (Capture)Suitable for: Sandwich ELISAReacts with: Human
Catalog No. NC2243551