Learn More
BPS Bioscience Inc Recombinant Cell Line, CHO, Human CD38, High Expression, 2 vials, ~2 x 10^6 cells/vial, Research Use Only
SDP
Catalog No. NC3779528
Encompass
This CD38 CHO recombinant cell line is designed for research applications focusing on CD38-mediated cellular processes and immunotherapy, constitutively expressing full-length human CD38 protein for studying CD38 biology, screening inhibitors, and developing therapies. This high expression level mimics cancer target cells, providing a relevant model for drug discovery and functional assays.
  • Physiologically relevant model for studying glycoprotein and ectoenzyme roles
  • High-expression model for immunotherapy target antigen investigations
  • Confirmed absence of mycoplasma contamination
  • Supplied as two vials with approx. 2 x 10^6 cells per vial
  • Storage buffer contains 10% DMSO in FBS
Catalog No. NC3779528 Supplier BPS Bioscience Inc Supplier No. 79615H
Add to Cart
Add to Cart
Request a Quote
This CD38 CHO recombinant cell line is designed for research applications focusing on CD38-mediated cellular processes and immunotherapy, constitutively expressing full-length human CD38 protein for studying CD38 biology, screening inhibitors, and developing therapies. This high expression level mimics cancer target cells, providing a relevant model for drug discovery and functional assays.
  • Physiologically relevant model for studying glycoprotein and ectoenzyme roles
  • High-expression model for immunotherapy target antigen investigations
  • Confirmed absence of mycoplasma contamination
  • Supplied as two vials with approx. 2 x 10^6 cells per vial
  • Storage buffer contains 10% DMSO in FBS