Learn More
ACROBiosystems Recombinant Protein;Human PCSK9 Protein;HEK293;50UG;PC9,PCSK9,PCSK-9,FH3,HCHOLA3,LDLCQ1,NARC-1,NARC1,PC9
Catalog No. NC0584518
Encompass_Preferred
Recombinant Protein;Human PCSK9 Protein;50UG;Human PCSK9, His Tag (PC9-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # AAI66619).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 75.1 kDa. The protein migrates as 20 kDa and 62 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation and proteolytic digestion.;Immobilized Human LDL R, His Tag (Cat. No. LDR-H5224) at 10 μg/mL (100 μL/well) can bind Human PCSK9, His Tag (Cat. No. PC9-H5223) with a linear range of 0.031-0.125 μg/mL (QC tested).
Catalog No. NC0584518 Supplier ACROBiosystems Supplier No. PC9H522350UG
Recombinant Protein;Human PCSK9 Protein;50UG;Human PCSK9, His Tag (PC9-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # AAI66619).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 75.1 kDa. The protein migrates as 20 kDa and 62 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation and proteolytic digestion.;Immobilized Human LDL R, His Tag (Cat. No. LDR-H5224) at 10 μg/mL (100 μL/well) can bind Human PCSK9, His Tag (Cat. No. PC9-H5223) with a linear range of 0.031-0.125 μg/mL (QC tested).