Learn More
Abcam Recombinant human PLK3 protein, 10UG
Supplier:  Abcam AB12562710UG
Encompass_Preferred
Recombinant human PLK3 protein is a Human Fragment protein, in the 57 to 340 aa range, expressed in Baculovirus infected Sf9, with >75% purity and suitable for SDS-PAGE, WB, FuncS.
Protein/ Peptide/ Enzyme Synonyms: CNK, FNK, PRK, PLK3, Serine/threonine-protein kinase PLK3, Cytokine-inducible serine/threonine-protein kinase, FGF-inducible kinase, Polo-like kinase 3, Proliferation-related kinase, PLK-3
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2063241