Learn More
Abcam Recombinant Human PNPLA3 protein, 10UG
Supplier:  Abcam AB13278710UG
Encompass_Preferred
Recombinant Human PNPLA3 protein is a Human Full Length protein, in the 1 to 481 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: ADPN, C22orf20, PNPLA3, 1-acylglycerol-3-phosphate O-acyltransferase PNPLA3, Acylglycerol transacylase, Adiponutrin, Calcium-independent phospholipase A2-epsilon, Lysophosphatidic acid acyltransferase, Patatin-like phospholipase domain-containing protein 3, iPLA2-epsilon
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2028357