Learn More
Abcam Recombinant Human PTPN22 protein, 10UG
Supplier:  Abcam AB13205510UG
Encompass_Preferred
Recombinant Human PTPN22 protein is a Human Full Length protein, in the 1 to 179 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: PTPN8, PTPN22, Tyrosine-protein phosphatase non-receptor type 22, Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP, Lymphoid phosphatase, PEST-domain phosphatase, LyP, PEP
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2169682