Learn More
Abcam Recombinant Human Tau (biotinylated ) protein
Supplier:  Abcam AB26901820UG
Encompass_Preferred
Recombinant Human Tau (biotinylated ) protein is a Full Length protein, in the 1 to 441 aa range, expressed in Escherichia coli, with >80% purity and suitable for SDS-PAGE.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2255194