Learn More
Abcam Recombinant Human/Murine/Rat BDNF protein (Active), 10UG
Supplier:  Abcam AB979410UG
Encompass_Preferred
Recombinant Human/Murine/Rat BDNF protein (Active) is a Human Full Length protein, in the 129 to 247 aa range, expressed in Escherichia coli, with >98% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, HPLC, WB, CellAct.
Protein/ Peptide/ Enzyme Synonyms: Neurotrophic factor BDNF precursor form, proBDNF, Abrineurin, Brain-derived neurotrophic factor, BDNF
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2256518