Learn More
Abcam Recombinant Human Apolipoprotein E, 500UG
Supplier:  Abcam AB55210500UG
Encompass_Preferred
Recombinant Human Apolipoprotein E is a Human Full Length protein with >90% purity and suitable for western blot, SDS-PAGE and HPLC.The predicted molecular weight of ab55210 protein is 34.3 kDa. - Save time and ensure accurate results - use our ApoE protein as a control
Protein/ Peptide/ Enzyme Synonyms: Apolipoprotein E, Apo-E, APOE
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1362058