Learn More
Abcam Recombinant Human Galectin 3 protein (His tag), 200UG
Supplier:  Abcam AB179972200UG
Encompass_Preferred
Recombinant Human Galectin 3 protein (His tag) is a Human Full Length protein, in the 1 to 250 aa range, expressed in Escherichia coli, with >92% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: MAC2, LGALS3, Galectin-3, Gal-3, 35 kDa lectin, Carbohydrate-binding protein 35, Galactose-specific lectin 3, Galactoside-binding protein, IgE-binding protein, L-31, Laminin-binding protein, Lectin L-29, Mac-2 antigen, CBP 35, GALBP
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2052164