Learn More
Abcam Recombinant human HPRT protein (Active), 20UG
Supplier:  Abcam AB22295120UG
Encompass_Preferred
Recombinant human HPRT protein (Active) is a Human Full Length protein, in the 1 to 218 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS, MS.
Protein/ Peptide/ Enzyme Synonyms: HPRT, HPRT1, Hypoxanthine-guanine phosphoribosyltransferase, HGPRT, HGPRTase
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2254609