Learn More
Abcam Recombinant human JAK3 protein, 10UG
Supplier:  Abcam AB8975610UG
Encompass_Preferred
Recombinant human JAK3 protein is a Human Fragment protein, in the 781 to 1124 aa range, expressed in Baculovirus infected Sf9, with >80% purity and suitable for SDS-PAGE, WB, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Tyrosine-protein kinase JAK3, Janus kinase 3, Leukocyte janus kinase, JAK-3, L-JAK, JAK3
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2066032