Learn More
Proteintech Group Inc Recomb. Human Activin A 1000ug
SDP
Supplier:  Proteintech Group Inc HZ11381000UG
Encompass_Preferred
Animal-free Recombinant Human Activin A is a member of the TGF beta superfamily, expressed from human 293 cells as disulfide linked homodimer with an apparent molecular mass of 25 kDa. This cytokine is produced in a serum-free, chemically defined media.
Catalog No. 50-197-3905
Add to Cart
Request a Quote