Learn More
Proteintech Group Inc Recomb. Human beta NGF 100ug
SDP
Supplier:  Proteintech Group Inc HZ1222100UG
Encompass_Preferred
Animal-free Recombinant Human beta NGF is expressed in human 293 cells as a non-disulfide bonded homodimeric protein with an apparent molecular mass of 13 kDa. Beta NGF is critical for the survival and maintenance of sympathetic and sensory neurons and may play an important role in the regulation of the immune system. The presence of beta NGF in immune cells and endocrine cells as well as in the CNS limbic areas suggests that beta NGF may function as an intracellular messenger to regulate the body’s response to stress. This product is produced in a human cell expression system with serum-free, chemically defined media.
Catalog No. 50-197-3934